siRNA Details
Gene Symbol: Akirin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007589.3 | NP_001007590.2 | 2 | 744 | ||
GenBank mRNA | AK005836.1 | - | 2 | 423 | ||
AK215036.1 | - | 1 | 98 | |||
AK217021.1 | - | 2 | 108 | |||
BC082305.1 | - | 3 | 432 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
