siRNA Details
Gene Symbol: Med18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026039.3 | NP_080315.1 | 3 | 348 | ||
GenBank mRNA | AK012886.1 | - | 3 | 349 | ||
AK012903.1 | - | 3 | 325 | |||
AK159696.1 | - | 3 | 351 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
