siRNA Details
Gene Symbol: Prpf18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026045.3 | NP_080321.2 | 10 | 2892 | ||
GenBank mRNA | AK013280.1 | - | 1 | 700 | ||
AK028921.1 | - | 3 | 2029 | |||
AK135707.1 | - | 10 | 2933 | |||
AK135890.1 | - | 10 | 2915 | |||
AK161550.1 | - | 6 | 2336 | |||
BC004573.1 | - | 10 | 2892 | 3586127 | ||
BC019370.1 | - | 1 | 1167 | |||
BC028306.1 | - | 10 | 2892 | |||
BC046822.1 | - | 1 | 605 | |||
BC058631.1 | - | 10 | 2887 | 6402639 |
Gene Symbol: 2610005L07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC058969.1 | - | 6 | 3992 | 6825946 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK162785.1 | - | 1760 | Chr. - - [-] on Build |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
