siRNA Details
Gene Symbol: Taok1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144825.2 | NP_659074.2 | 20 | 10984 | ||
GenBank mRNA | AK017320.1 | - | 1 | 785 | ||
AK173156.1 | - | 4 | 8319 | |||
BC019960.1 | - | 1 | 1029 | |||
BC092076.1 | - | 1 | 1095 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
