siRNA Details
Gene Symbol: Herc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026101.4 | NP_080377.2 | 3 | 589 | ||
NM_030114.2 | NP_084390.1 | 3 | 589 | |||
GenBank mRNA | AK009436.1 | - | 3 | 444 | ||
AK019546.1 | - | 3 | 583 | |||
AK143111.1 | - | 3 | 463 | |||
AK147723.1 | - | 3 | 476 | |||
BC043082.1 | - | 3 | 401 | 6417111 | ||
BC060033.1 | - | 1 | 174 | 6478123 | ||
BQ899503.1 | - | 3 | 590 |
