siRNA Details
Gene Symbol: Akirin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007589.3 | NP_001007590.2 | 5 | 1147 | ||
GenBank mRNA | AK004133.1 | - | 1 | 92 | ||
AK005836.1 | - | 5 | 829 | |||
BC082305.1 | - | 5 | 3507 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
