siRNA Details
Gene Symbol: Eif2s2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026030.2 | NP_080306.1 | 3, 4 | 507 | ||
GenBank mRNA | AK012338.1 | - | 3, 4 | 420 | ||
AK012817.1 | - | 2, 3 | 297 | |||
AK076056.1 | - | 3, 4 | 427 | |||
AK145401.1 | - | 3, 4 | 412 | |||
AK145434.1 | - | 3, 4 | 454 | |||
AK166862.1 | - | 3, 4 | 411 | |||
BC003848.1 | - | 3, 4 | 383 | 3594058 | ||
BC100539.1 | - | 3, 4 | 418 | |||
BC113766.1 | - | 3, 4 | 379 | 40058503 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
