siRNA Details
Gene Symbol: 3110040N11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026077.3 | NP_080353.1 | 3, 4 | 362 | ||
GenBank mRNA | AK014163.1 | - | 3, 4 | 342 | ||
AK018003.1 | - | 3, 4 | 388 | |||
AK019261.1 | - | 3, 4 | 362 | |||
AK170168.1 | - | 3, 4 | 379 | |||
BC027500.1 | - | 3, 4 | 350 | 1448876 |
