siRNA Details
Gene Symbol: Atp8b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006684.1 | - | 3, 4 | 261 | ||
AK018868.1 | - | 3, 4 | 261 | |||
AK018877.1 | - | 3, 4 | 261 | |||
BC049570.1 | - | 3, 4 | 231 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
