siRNA Details
Gene Symbol: 1700093K21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110133.1 | NP_001103603.1 | 3, 4 | 752 | ||
NM_026105.3 | NP_080381.2 | 3, 4 | 434 | |||
GenBank mRNA | AK007052.1 | - | 3, 4 | 340 | ||
AK007139.1 | - | 2, 3 | 191 | |||
BC049567.1 | - | 3, 4 | 434 | 6773879 |
