siRNA Details
Gene Symbol: Paxbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026110.2 | NP_080386.3 | 11 | 1881 | ||
GenBank mRNA | AK007365.1 | - | 5 | 586 | ||
AK079394.1 | - | 4 | 746 | |||
AY033907.1 | - | 11 | 1666 | |||
AY033908.1 | - | 11 | 1683 | |||
BC014838.1 | - | 4 | 366 | 3588322 | ||
BC027145.1 | - | 3 | 301 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
