siRNA Details
Gene Symbol: Hmgn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026122.4 | NP_080398.1 | 5 | 386 | ||
GenBank mRNA | AK002970.1 | - | 5 | 305 | ||
AK153223.1 | - | 5 | 346 | |||
BC005693.1 | - | 5 | 322 | 3597594 | ||
CJ150650.1 | - | 5 | 383 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
