siRNA Details
Gene Symbol: TAZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000116.4 | NP_000107.1 | 6, 7 | 875 | ||
NM_181311.3 | NP_851828.1 | 5, 6 | 785 | |||
NR_024048.2 | 5, 6 | 891 | ||||
XM_006724836.1 | XP_006724899.1 | 6, 7 | 906 | |||
XM_006724841.3 | XP_006724904.1 | 6, 7 | 627 | |||
XM_006724842.3 | XP_006724905.1 | 5, 6 | 537 | |||
XM_011531191.2 | XP_011529493.1 | 5, 6 | 1065 | |||
XM_017029762.1 | XP_016885251.1 | 5, 6 | 816 | |||
XM_017029764.1 | XP_016885253.1 | 4, 5 | 404 | |||
GenBank mRNA | AK291848.1 | - | 6, 7 | 743 | ||
AY231461.1 | - | 5, 6 | 467 | |||
AY231464.1 | - | 6, 7 | 557 | |||
AY258036.1 | - | 6, 7 | 557 | |||
AY258039.1 | - | 5, 6 | 467 | |||
BC005062.1 | - | 5, 6 | 848 | |||
KU178349.1 | - | 6, 7 | 549 | |||
KU178352.1 | - | 5, 6 | 459 | |||
X92762.1 | - | 6, 7 | 837 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
