siRNA Details
Gene Symbol: 1700001G17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005592.1 | - | 1 | 555 | ||
AK008236.1 | - | 1 | 538 | |||
AK008325.1 | - | 1 | 538 | |||
BC132564.1 | - | 1 | 358 | |||
BC132566.1 | - | 1 | 358 |
