siRNA Details
Gene Symbol: Paxbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026110.2 | NP_080386.3 | 17 | 2563 | ||
GenBank mRNA | AK007365.1 | - | 11 | 1268 | ||
AK011477.1 | - | 7 | 647 | |||
AY033907.1 | - | 17 | 2348 | |||
AY033908.1 | - | 17 | 2365 | |||
BC014838.1 | - | 10 | 1048 | 3588322 | ||
BC027145.1 | - | 9 | 983 |

More Information
Other Species Targeted

Gene Ontology Categories:
