siRNA Details
Gene Symbol: Tmem167b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026198.2 | NP_080474.2 | 3 | 1792 | ||
GenBank mRNA | AK008415.1 | - | 3 | 1706 | ||
AK046697.1 | - | 3 | 1692 | |||
AK155399.1 | - | 3 | 1688 | |||
BC050932.1 | - | 3 | 1696 | 6811263 | ||
BC055449.1 | - | 3 | 1714 | 3993993 |
