siRNA Details
Gene Symbol: Cpeb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290676.1 | NP_001277605.1 | 5 | 2847 | ||
NM_001290678.1 | NP_001277607.1 | 4 | 2796 | |||
NM_026252.4 | NP_080528.2 | 6 | 2871 | |||
GenBank mRNA | AK015381.1 | - | 3 | 266 | ||
AK015401.1 | - | 2 | 254 | |||
BC079599.1 | - | 5 | 655 | |||
BC115430.1 | - | 4 | 1520 | 40094123 | ||
BC115431.1 | - | 4 | 1520 | 40094124 | ||
BC145863.1 | - | 6 | 1595 | 40131166 | ||
BC145865.1 | - | 6 | 1595 | 40131168 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
