siRNA Details
Gene Symbol: 2010106E10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168590.1 | NP_001162062.1 | 2 | 205 | ||
NM_026333.4 | NP_080609.2 | 2 | 205 | |||
GenBank mRNA | AK008319.1 | - | 2 | 206 | ||
AK017156.1 | - | 2 | 207 | |||
BC139480.1 | - | 2 | 162 | 8862502 | ||
BC139481.1 | - | 2 | 162 | 8862503 |
