siRNA Details
Gene Symbol: Alg13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026247.3 | NP_080523.2 | 2 | 275 | ||
GenBank mRNA | AK008162.1 | - | 2 | 225 | ||
AK012910.1 | - | 287 | ||||
AK014790.1 | - | 2 | 220 | |||
AK086546.1 | - | 2 | 168 | |||
AK089102.1 | - | 2 | 239 | |||
BC053004.1 | - | 2 | 212 | |||
BC137692.1 | - | 2 | 275 | 8860711 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
