siRNA Details
Gene Symbol: GALC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000153.3 | NP_000144.2 | 17 | 2919 | ||
NM_001201401.1 | NP_001188330.1 | 16 | 2850 | |||
NM_001201402.1 | NP_001188331.1 | 17 | 2900 | |||
XM_011536618.2 | XP_011534920.1 | 17 | 3201 | |||
GenBank mRNA | AK302519.1 | - | 12 | 1456 | ||
AK302683.1 | - | 17 | 2194 | |||
BC036518.2 | - | 17 | 2772 | 5267705 | ||
D25283.1 | - | 17 | 2802 | |||
L23116.1 | - | 17 | 2818 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
