siRNA Details
Gene Symbol: Cep83os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016525.1 | - | 3 | 2235 | ||
AK144701.1 | - | 3 | 2238 | |||
AK154718.1 | - | 3 | 2208 | |||
AK155868.1 | - | 3 | 2208 | |||
BC021358.1 | - | 1 | 1053 | |||
BC061482.1 | - | 1 | 1251 | |||
BC067070.1 | - | 3 | 2221 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
