siRNA Details
Gene Symbol: Spaca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290443.1 | NP_001277372.1 | 3 | 417 | ||
NM_026293.3 | NP_080569.1 | 3 | 419 | |||
GenBank mRNA | AK006187.1 | - | 3 | 420 | ||
AK007270.1 | - | 3 | 391 | |||
AK016011.1 | - | 3 | 412 | |||
BC147079.1 | - | 3 | 416 | 9056040 | ||
BC147080.1 | - | 3 | 416 | 9056039 | ||
BY706285.1 | - | 3 | 418 |
