siRNA Details
Gene Symbol: Dnajc19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286973.1 | NP_001273902.1 | 4, 5 | 401 | ||
NM_026332.4 | NP_080608.3 | 5, 6 | 453 | |||
GenBank mRNA | AK007866.1 | - | 5, 6 | 421 | ||
AK010204.1 | - | 5, 6 | 403 | |||
AK012051.1 | - | 5, 6 | 405 | |||
BC020327.1 | - | 5, 6 | 386 | |||
CA469052.1 | - | 5, 6 | 490 | |||
CJ123658.1 | - | 4, 5 | 358 |

More Information
Other Species Targeted

Gene Ontology Categories:
