siRNA Details
Gene Symbol: Ppid | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026352.3 | NP_080628.1 | 2 | 252 | ||
GenBank mRNA | AK003402.1 | - | 2 | 209 | ||
AK013919.1 | - | 2 | 243 | |||
AK016221.1 | - | 2 | 248 | |||
AK051597.1 | - | 2 | 232 | |||
AK142413.1 | - | 2 | 300 | |||
AK150520.1 | - | 2 | 269 | |||
AK151091.1 | - | 2 | 269 | |||
BC011499.1 | - | 2 | 249 | 3584183 | ||
BC019778.1 | - | 2 | 252 | 4035785 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
