siRNA Details
Gene Symbol: Slc48a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026353.4 | NP_080629.1 | 3 | 746 | ||
GenBank mRNA | AB041659.1 | - | 3 | 1944 | ||
AK002729.1 | - | 3 | 689 | |||
AK007888.1 | - | 3 | 695 | |||
AK088485.1 | - | 3 | 669 | |||
AK138729.1 | - | 3 | 682 | |||
AK150552.1 | - | 3 | 695 | |||
AK155824.1 | - | 3 | 733 | |||
AK159879.1 | - | 3 | 712 | |||
AK159898.1 | - | 3 | 665 | |||
AK168636.1 | - | 3 | 678 | |||
AK170393.1 | - | 3 | 723 | |||
AK181240.1 | - | 1 | 219 | |||
AK184893.1 | - | 1 | 218 | |||
AK189427.1 | - | 1 | 154 | |||
AK189571.1 | - | 1 | 155 | |||
AK193857.1 | - | 1 | 217 | |||
AK204097.1 | - | 1 | 94 | |||
AK213892.1 | - | 1 | 218 | |||
AK219716.1 | - | 1 | 89 | |||
BC022913.1 | - | 3 | 692 | 4237364 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
