siRNA Details
Gene Symbol: Zfp169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164575.1 | NP_001158047.1 | 5 | 1486 | ||
NM_026450.3 | NP_080726.3 | 5 | 1518 | |||
GenBank mRNA | AK006349.1 | - | 1 | 581 | ||
AK016063.1 | - | 1 | 570 | |||
BC060222.1 | - | 5 | 1476 | 30356624 |
