siRNA Details
Gene Symbol: 1700129C05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026461.2 | NP_080737.1 | 1 | 242 | ||
GenBank mRNA | AK006084.1 | - | 1 | 244 | ||
AK007313.1 | - | 1 | 242 | |||
AK015075.1 | - | 1 | 229 | |||
BC049552.1 | - | 1 | 242 | 6774189 |
Gene Symbol: Gm5142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004158.2 | NP_001004158.1 | 1 | 232 | ||
GenBank mRNA | AK076969.1 | - | 1 | 233 | ||
BC099536.1 | - | 1 | 204 | 30918677 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
