siRNA Details
Gene Symbol: Asprv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026414.2 | NP_080690.2 | 1 | 962 | ||
GenBank mRNA | AK004007.1 | - | 1 | 409 | ||
AK160494.1 | - | 1 | 903 | |||
BC057938.1 | - | 1 | 975 | |||
BC108357.1 | - | 1 | 860 | |||
BC119099.1 | - | 1 | 478 | |||
BC119101.1 | - | 1 | 478 | |||
DQ841260.1 | - | 1 | 962 |

More Information
Other Species Targeted

Gene Ontology Categories:
