siRNA Details
Gene Symbol: Hcfc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081218.1 | NP_001074687.1 | 13 | 1855 | ||
GenBank mRNA | AK007317.1 | - | 4 | 338 | ||
BC116411.1 | - | 6 | 647 | |||
BC127176.1 | - | 6 | 647 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
