siRNA Details
Gene Symbol: Lamp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177356.3 | NP_796330.2 | 6 | 2618 | ||
GenBank mRNA | AJ510130.1 | - | 6 | 2529 | ||
AK028039.1 | - | 6 | 2570 | |||
AK033379.1 | - | 6 | 2571 |
Gene Symbol: Ppid | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026352.3 | NP_080628.1 | 5 | 713 | ||
GenBank mRNA | AK003402.1 | - | 5 | 670 | ||
AK013919.1 | - | 5 | 704 | |||
AK016221.1 | - | 5 | 709 | |||
AK051597.1 | - | 5 | 693 | |||
AK142413.1 | - | 4 | 3137 | |||
AK150520.1 | - | 5 | 730 | |||
AK151091.1 | - | 5 | 730 | |||
BC011499.1 | - | 5 | 710 | 3584183 | ||
BC019778.1 | - | 5 | 713 | 4035785 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
