siRNA Details
Gene Symbol: LAMA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000227.4 | NP_000218.3 | 1 | 134 | ||
NM_001127718.2 | NP_001121190.2 | 1 | 134 | |||
GenBank mRNA | AY327114.1 | - | 1 | 109 | ||
DA862923.1 | - | 1 | 134 | |||
L34155.1 | - | 1 | 76 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
