siRNA Details
Gene Symbol: Tmem39a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205286.1 | NP_001192215.1 | 9 | 1626 | ||
NM_001205287.1 | NP_001192216.1 | 9 | 1674 | |||
NM_026407.3 | NP_080683.2 | 9 | 1878 | |||
GenBank mRNA | AK011646.1 | - | 9 | 1855 | ||
AK017817.1 | - | 9 | 1836 | |||
AK133638.1 | - | 9 | 1850 | |||
AK160576.1 | - | 9 | 1627 | |||
AK169127.1 | - | 9 | 1676 | |||
BC020318.2 | - | 9 | 1833 | 5250532 |
