siRNA Details
Gene Symbol: Ssxb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026492.3 | NP_080768.1 | 2, 3 | 258 | ||
GenBank mRNA | AK006218.1 | - | 2, 3 | 259 | ||
AK015135.1 | - | 2, 3 | 259 | |||
AY244792.1 | - | 2, 3 | 259 | |||
BC145538.1 | - | 2, 3 | 231 | 9054101 | ||
BC145541.1 | - | 176 | 9054104 | |||
BC145782.1 | - | 2, 3 | 150 | 40131085 | ||
BC145784.1 | - | 2, 3 | 150 | 40131087 | ||
BC171964.1 | - | 231 | 9054250 |
Gene Symbol: Ssxb12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY244803.1 | - | 1, 2 | 122 |
Gene Symbol: Ssxb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205108.1 | NP_001192037.1 | 82 | |||
GenBank mRNA | AY244797.1 | - | 83 |
Gene Symbol: Ssxb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY244798.1 | - | 83 |
Gene Symbol: Ssxb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY244799.1 | - | 83 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
