siRNA Details
Gene Symbol: Cspp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026493.3 | NP_080769.3 | 3 | 301 | ||
GenBank mRNA | AK013065.1 | - | 3 | 270 | ||
AK015133.1 | - | 3 | 270 | |||
AK032310.1 | - | 3 | 281 | |||
AK077173.1 | - | 3 | 289 | |||
BC019581.1 | - | 3 | 257 | |||
BC151042.1 | - | 3 | 274 | 9087955 | ||
CN459376.1 | - | 3 | 271 | |||
FJ008676.1 | - | 2 | 128 | |||
FJ008677.1 | - | 2 | 164 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
