siRNA Details
Gene Symbol: MAP3K12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193511.1 | NP_001180440.1 | 14 | 3422 | ||
NM_006301.3 | NP_006292.3 | 15 | 3323 | |||
XM_005269138.3 | XP_005269195.1 | 14 | 3350 | |||
XM_006719588.3 | XP_006719651.1 | 14 | 3244 | |||
XM_011538725.2 | XP_011537027.1 | 14 | 4227 | |||
XM_017019956.1 | XP_016875445.1 | 15 | 3251 | |||
GenBank mRNA | AB209453.1 | - | 5 | 5407 | ||
BC050050.1 | - | 11 | 2303 | 4824512 | ||
U07358.1 | - | 15 | 3150 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
