siRNA Details
Gene Symbol: Ssxb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026492.3 | NP_080768.1 | 3 | 267 | ||
GenBank mRNA | AK006218.1 | - | 3 | 268 | ||
AK015135.1 | - | 3 | 268 | |||
AY244792.1 | - | 3 | 268 | |||
BC145538.1 | - | 3 | 240 | 9054101 | ||
BC145541.1 | - | 185 | 9054104 | |||
BC145782.1 | - | 3 | 159 | 40131085 | ||
BC145784.1 | - | 3 | 159 | 40131087 | ||
BC171964.1 | - | 240 | 9054250 |
Gene Symbol: Gm14459 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126491.1 | NP_001119963.1 | 2 | 113 |
Gene Symbol: Gm6592 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081564.1 | NP_001075033.1 | 2 | 86 |
Gene Symbol: Ssxb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199064.1 | NP_951019.1 | 3 | 268 | ||
GenBank mRNA | AY244801.1 | - | 3 | 268 | ||
BC145539.1 | - | 240 | 9054102 |
Gene Symbol: Ssxb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY244802.1 | - | 2 | 131 |
Gene Symbol: Ssxb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001450.4 | NP_001001450.1 | 4 | 444 | ||
NM_001134226.1 | NP_001127698.1 | 3 | 267 | |||
GenBank mRNA | AK076879.1 | - | 4 | 445 | ||
AY244793.1 | - | 4 | 445 | |||
BC048441.1 | - | 3 | 267 | 6773922 | ||
BC089478.1 | - | 3 | 267 | 6743842 |
Gene Symbol: Ssxb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198898.2 | NP_942601.1 | 2 | 132 | ||
GenBank mRNA | AY244794.1 | - | 2 | 132 | ||
BC087939.1 | - | 3 | 212 | |||
BC132618.1 | - | 2 | 132 | 40130895 | ||
BC132644.1 | - | 2 | 132 | 40130921 |
Gene Symbol: Ssxb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199319.3 | NP_955023.1 | 2 | 131 | ||
GenBank mRNA | AY244796.1 | - | 2 | 131 |
Gene Symbol: Ssxb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205108.1 | NP_001192037.1 | 91 | |||
GenBank mRNA | AY244797.1 | - | 92 |
Gene Symbol: Ssxb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY244798.1 | - | 92 |
Gene Symbol: Ssxb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081565.3 | NP_001075034.3 | 2 | 91 | ||
GenBank mRNA | AY244799.1 | - | 92 |
Gene Symbol: Ssxb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199066.2 | NP_951021.2 | 3 | 268 | ||
GenBank mRNA | AY244800.1 | - | 268 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
