siRNA Details
Gene Symbol: N6amt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026526.2 | NP_080802.1 | 4 | 495 | ||
GenBank mRNA | AK005312.1 | - | 4 | 489 | ||
AK010911.1 | - | 3 | 490 | |||
AK012083.1 | - | 4 | 496 | |||
BC051925.1 | - | 4 | 486 | 5691329 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
