siRNA Details
Gene Symbol: Dcakd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026551.3 | NP_080827.2 | 3 | 492 | ||
GenBank mRNA | AK051022.1 | - | 3 | 493 | ||
AK078460.1 | - | 3 | 425 | |||
AK162069.1 | - | 3 | 452 | |||
AK170762.1 | - | 3 | 443 | |||
AK190020.1 | - | 1 | 189 | |||
BC005607.1 | - | 3 | 458 | 3498151 | ||
BC023208.1 | - | 2 | 406 | |||
BC031855.1 | - | 3 | 441 | |||
BC042685.1 | - | 2 | 406 | |||
BC045193.1 | - | 3 | 446 | 5368157 | ||
BC047340.1 | - | 2 | 406 |
