siRNA Details
Gene Symbol: D730048I06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026593.3 | NP_080869.1 | 3 | 737 | ||
GenBank mRNA | AK020302.1 | - | 3 | 709 | ||
AK020310.1 | - | 3 | 690 | |||
AK021351.1 | - | 3 | 709 | |||
AK033813.1 | - | 3 | 738 | |||
AK136636.1 | - | 3 | 708 |
