siRNA Details
Gene Symbol: Atad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026487.3 | NP_080763.2 | 10 | 1923 | ||
GenBank mRNA | AK009419.1 | - | 6 | 1511 | ||
AK014967.1 | - | 10 | 1940 | |||
AK018102.1 | - | 1 | 782 | |||
AK030719.1 | - | 10 | 1944 | |||
AK033639.1 | - | 10 | 1947 | |||
AK150469.1 | - | 10 | 1943 | |||
AK152059.1 | - | 10 | 1943 | |||
AK165953.1 | - | 10 | 1958 | |||
AK210281.1 | - | 1 | 34 | |||
BC043051.1 | - | 10 | 1908 | 5708377 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
