siRNA Details
Gene Symbol: Cdc42ep3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026514.2 | NP_080790.1 | 2 | 1681 | ||
GenBank mRNA | AK014281.1 | - | 2 | 1683 | ||
AK019447.1 | - | 2 | 1683 | |||
AK076782.1 | - | 2 | 1480 | |||
AK150214.1 | - | 2 | 1648 |
