siRNA Details
Gene Symbol: Cep57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026665.4 | NP_080941.3 | 2 | 283 | ||
GenBank mRNA | AK014873.1 | - | 2 | 230 | ||
AK016484.1 | - | 2 | 67 | |||
AK019414.1 | - | 2 | 165 | |||
AK028408.1 | - | 2 | 132 | |||
AK028458.1 | - | 2 | 111 | |||
AK129053.1 | - | 2 | 232 | |||
AK167523.1 | - | 2 | 168 | |||
AY225093.1 | - | 2 | 76 | |||
BB846596.1 | - | 2 | 283 | |||
BC025588.1 | - | 1 | 63 | |||
BC030353.1 | - | 2 | 554 | |||
BC056968.1 | - | 2 | 529 | |||
BY134402.1 | - | 2 | 270 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
