siRNA Details
Gene Symbol: PHKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000293.2 | NP_000284.1 | 1, 2 | 136 | ||
XM_005255983.4 | XP_005256040.1 | 1, 2 | 99 | |||
XM_017023284.1 | XP_016878773.1 | 1, 2 | 102 | |||
GenBank mRNA | AK309226.1 | - | 1, 2 | 99 | ||
X84908.1 | - | 1, 2 | 108 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
