siRNA Details
Gene Symbol: Blvra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026678.4 | NP_080954.4 | 3 | 273 | ||
GenBank mRNA | AK002231.1 | - | 2 | 212 | ||
AK010847.1 | - | 3 | 228 | |||
AK045022.1 | - | 3 | 266 | |||
AK162186.1 | - | 3 | 234 | |||
AK172620.1 | - | 3 | 238 | |||
BY073963.1 | - | 3 | 274 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
