siRNA Details
Gene Symbol: PLOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000302.3 | NP_000293.2 | 1, 2 | 196 | ||
GenBank mRNA | AK294688.1 | - | 1, 2 | 138 | ||
AK295254.1 | - | 1, 2 | 140 | |||
AU139472.1 | - | 1, 2 | 175 | |||
BC016657.2 | - | 1, 2 | 100 | 3917351 | ||
DC334246.1 | - | 1, 2 | 196 | |||
M98252.1 | - | 1, 2 | 107 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
