siRNA Details
Gene Symbol: 0610010K14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177601.1 | NP_001171072.1 | 4, 5 | 520 | ||
NM_026757.2 | NP_081033.1 | 3, 4 | 418 | |||
NM_145758.1 | NP_665701.1 | 4, 5 | 520 | |||
GenBank mRNA | AA189586.1 | - | 1, 2 | 26 | ||
AK003842.1 | - | 3, 4 | 418 | |||
AK208696.1 | - | 248 | ||||
CK791552.1 | - | 4, 5 | 499 | |||
EL959640.1 | - | 4, 5 | 465 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001127521, XM_006246652, XM_006246655, XM_006246653, BC158879, and others.
Gene Ontology Categories:
