siRNA Details
Gene Symbol: Fam166a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026624.3 | NP_080900.1 | 7 | 1104 | ||
GenBank mRNA | AK016459.1 | - | 7 | 1106 | ||
BC061039.1 | - | 7 | 1107 | 6704633 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
