siRNA Details
Gene Symbol: Pomgnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290658.1 | NP_001277587.1 | 15 | 1442 | ||
NM_026651.2 | NP_080927.1 | 16 | 1528 | |||
NM_029786.2 | NP_084062.2 | 15 | 1429 | |||
GenBank mRNA | AB053220.1 | - | 16 | 1510 | ||
AK002638.1 | - | 15 | 1421 | |||
AK015478.1 | - | 15 | 1444 | |||
AK019640.1 | - | 15 | 1434 | |||
AK177045.1 | - | 3 | 219 | |||
AK212416.1 | - | 1 | 50 | |||
BC011201.1 | - | 16 | 1451 | 4190368 | ||
BC023265.1 | - | 13 | 1116 |
