siRNA Details
Gene Symbol: Dopey2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026700.2 | NP_080976.1 | 4 | 686 | ||
GenBank mRNA | AK002808.1 | - | 4 | 616 | ||
AK156170.1 | - | 3 | 1293 | |||
AK164839.1 | - | 4 | 663 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
