siRNA Details
Gene Symbol: Ifitm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112715.1 | NP_001106186.1 | 2 | 563 | ||
NM_026820.3 | NP_081096.3 | 3 | 537 | |||
GenBank mRNA | AK004121.1 | - | 2 | 426 | ||
AK169960.1 | - | 2 | 408 | |||
AV136334.1 | - | 1 | 160 | |||
BC090258.1 | - | 2 | 391 | 4982747 | ||
BC090972.1 | - | 2 | 491 | 30297959 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
